MilliporeSigma
  • Automated online dual-column extraction coupled with teicoplanin stationary phase for simultaneous determination of (R)- and (S)-propranolol in rat plasma using liquid chromatography-tandem mass spectrometry.
Automated online dual-column extraction coupled with teicoplanin stationary phase for simultaneous determination of (R)- and (S)-propranolol in rat plasma using liquid chromatography-tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2003-04-23)
Yuan-Qing Xia, Ray Bakhtiar, Ronald B Franklin
ABSTRACT
An automated online sample extraction method for rat plasma was developed and validated for the quantification of (R)- and (S)-propranolol following the intravenous administration of either the racemate or the individual enantiomers at 5 mg/kg. A dual-column extraction system coupled to a chiral stationary phase (CSP) was used in conjunction with liquid chromatography-tandem mass spectrometry. In this method, two Oasis HLB extraction columns (50x1.0 mm) in parallel were used for online plasma sample purification and teicoplanin CSP (Chirobiotic T) was used for the enantiomeric separation. This method allowed the use of one of the extraction columns for purification while the other was being equilibrated. Hence, the time required for re-conditioning the extraction columns did not contribute to the total analysis time per sample, which resulted in a relatively shorter run time and higher throughput. The lower limit of detection was 0.5 ng/ml and the lower limit of quantification was 2 ng/ml for each enantiomer using 25 microl of rat plasma. The method was validated with a linear calibration curve between 2 and 2000 ng/ml for (R)- and (S)-propranolol, respectively. The intra- and inter-day precision (C.V.) was no more than 7.6% and the accuracy of the assay was between 92 and 103%. The teicoplanin CSP proved to be rugged with excellent reproducibility of chromatographic parameters.
Product Description
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Astec® CHIROBIOTIC® T Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm