MilliporeSigma
  • HPLC and 1H-NMR study of chiral recognition in some thromboxane antagonists induced by beta-cyclodextrin.
HPLC and 1H-NMR study of chiral recognition in some thromboxane antagonists induced by beta-cyclodextrin.
Journal of pharmaceutical and biomedical analysis (1991-01-01)
A F Casy, A D Cooper, T M Jefferies, R M Gaskell, D Greatbanks, R Pickford
ABSTRACT
The interaction of beta-cyclodextrin with a series of structurally related chiral thromboxane antagonists was investigated using NMR and RP-HPLC. HPLC studies used both a cyclodextrin bonded phase (Cyclobond I), and beta-cyclodextrin as a mobile phase additive with an achiral C8 column. Many of the compounds exhibited chiral recognition with beta-cyclodextrin in each technique, but only partial correlations between the three data sets were observed. HPLC and ROESY NMR data suggested the possibility of bimodal inclusion.
Product Description
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Astec® CYCLOBOND I 2000 Chiral HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm