MilliporeSigma
  • Palladium-catalyzed enantioselective alpha-arylation and alpha-vinylation of oxindoles facilitated by an axially chiral P-stereogenic ligand.
Palladium-catalyzed enantioselective alpha-arylation and alpha-vinylation of oxindoles facilitated by an axially chiral P-stereogenic ligand.
Journal of the American Chemical Society (2009-07-08)
Alexander M Taylor, Ryan A Altman, Stephen L Buchwald
ABSTRACT
The enantioselective alpha-arylation and alpha-vinylation of oxindoles catalyzed by Pd and a biarylmonophosphine ligand with both axial and phosphorus-based chirogenicity is reported. The resultant quaternary carbon stereocenters are formed in high enantiomeric excess, and the conditions tolerate a range of substitution on both the oxindole and the aryl/vinyl coupling partners.
Sigma-Aldrich
trans-1-Bromo-1-propene, contains copper as stabilizer, 99%