MilliporeSigma
(Z)-3-(1-Methyl-1H-indol-3-yl)-2-(thiophen-3-yl)acrylonitrile.
Acta crystallographica. Section C, Crystal structure communications (2004-03-09)
Vijayakumar N Sonar, Sean Parkin, Peter A Crooks
ABSTRACT
The title compound, C16H12N2S, has been synthesized by base-catalyzed condensation of 1-methylindole-3-carboxaldehyde with thiophene-3-acetonitrile. The product assumes an approximately planar Z configuration. The molecule has a thienyl-ring flip disorder.
Sigma-Aldrich
1-Methylindole-3-carboxaldehyde, 97%