MilliporeSigma
Fragrance material review on 4-tert-butylcyclohexyl acetate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-10-10)
S P Bhatia, L Jones, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of 4-tert-butylcyclohexyl acetate when used as a fragrance ingredient is presented.
Sigma-Aldrich
4-tert-Butylcyclohexyl acetate, mixture of cis and trans, 99%