MilliporeSigma
  • Nearly pure blue photoluminescent poly(2,7-[9-)3,5-bis[3,5- bis(benzyloxy)benzyloxy]benzyl)-9-(3,6-dioxaoctyl)]fluorene) in film.
Nearly pure blue photoluminescent poly(2,7-[9-)3,5-bis[3,5- bis(benzyloxy)benzyloxy]benzyl)-9-(3,6-dioxaoctyl)]fluorene) in film.
Chemical communications (Cambridge, England) (2002-09-25)
H Z Tang, M Fujiki, Z B Zhang, K Torimitsu, M Motonaga
ABSTRACT
The first unsymmetrically substituted polyfluorene bearing a bulky poly(benzyl ether) dendron and less bulky 3,6-dioxaoctyl groups in the 9-position was designed and synthesized, which gives almost a pure bluish photoluminescence with negligible weak greenish excimer emission around 520 nm even in a thermally annealed thin solid film.
Sigma-Aldrich
2,7-Dibromofluorene, 97%