MilliporeSigma
  • Tanshinone II A inhibits tat-induced HIV-1 transactivation through redox-regulated AMPK/Nampt pathway.
Tanshinone II A inhibits tat-induced HIV-1 transactivation through redox-regulated AMPK/Nampt pathway.
Journal of cellular physiology (2014-01-15)
Hong-Sheng Zhang, Xin-Yu Chen, Tong-Chao Wu, Feng-Juan Zhang
ABSTRACT
Tat transactivating activity regulated by NAD(+) -dependent histone deacetylase sirtuin1 (SIRT1) connects HIV transcription with the metabolic state of the cell. Nicotinamide phosphoribosyltransferase (Nampt) is a rate-limiting enzyme in the mammalian NAD(+) biosynthesis. Nampt, SIRT1, and AMPK were involved in inhibiting HIV-1 transactivation through redox-regulated pathway. Tanshinone II A is a main lipid-soluble monomer derivative from the root of Salvia miltiorrhiza (Danshen) and tanshinone II A possess a variety of biological activities through redox signaling pathway. Here we investigated the effect of tanshinone II A on Tat-induced HIV-1 transactivation and the redox signaling pathway involved in it. As the results were shown, tanshinone II A reversed Tat-induced reactive oxygen species (ROS) production and down-regulation of glutathione (GSH) levels in TZM-bl cells through up-regulation of Nrf2 expression. Tanshinone II A reversed Tat-induced inhibition of SIRT1 activity but not SIRT1 protein expression. Tanshinone II A reversed Tat-induced inhibition of Nampt protein expression and depletion of NAD(+) levels in TZM-bl cells in a dose-dependent manner. Tanshinone II A-evoked Nampt expression was mediated by AMPK signaling pathway. Tanshinone II A inhibited Tat-induced HIV-1 LTR transactivation dependent on AMPK-Nampt pathway. Collectively, our data provide new insights into understanding of the molecular mechanisms of tanshinone II A inhibited Tat-regulated transcription, suggesting that targeting AMPK/Nampt/SIRT1 pathway could serve as new anti-HIV-1 agents.
Sigma-Aldrich
Tanshinone IIA, ≥97% (HPLC)
Sigma-Aldrich
Tanshinone I, ≥98% (HPLC)
Tanshinone IIA, European Pharmacopoeia (EP) Reference Standard