MilliporeSigma
  • Self-assembly of supramolecualr metallogelator containing 2-(2'-hydroxyphenyl) benzoxazole/Zn(II) chelate.
Self-assembly of supramolecualr metallogelator containing 2-(2'-hydroxyphenyl) benzoxazole/Zn(II) chelate.
Journal of nanoscience and nanotechnology (2010-12-09)
Tae Hyeon Kim, Na Young Kwon, Ji Hye Son, Changduk Yang, Minjung Lee, Taek Seung Lee
ABSTRACT
A low molar mass organogelator 1 containing 2-(2'-hydroxyphenyl)benzoxazole (HPB) unit with long alkyl chain was synthesized by the reaction with HPB and octyl isocyanate in THF at room temperature. A new chelate-based organogelator 1-Zn(II) was prepared with the reaction of 1 and zinc(II) acetate in methanol and dichloromethane at mild condition. The gelation ability of organogelator was tested by heating-cooling method in various organic solvents, and the opaque gel was formed from DMF. Well-developed self-assembled structure of organogel was confirmed with field emission-scanning electron microscope (FE-SEM) and transmission electron microscope (TEM), and the optical properties of organogel upon aggregation were monitored by UV-Vis and fluorescence spectroscopy, 1-Zn(II) was self-assembled to the sheet-like structure with the thickness of fully extended length of molecules.
Sigma-Aldrich
Octyl isocyanate, 97%