MilliporeSigma
Potassium and magnesium retaining triamterene derivatives.
Die Pharmazie (1997-03-01)
H Priewer, F Ullrich
ABSTRACT
This work represents a summary of studies carried out with potassium and magnesium retaining triamterene derivatives. The synthesis, analytics and physico-chemical characteristics of those substances are described. Furthermore, the relationships between the chemical structure and electrolyte excretion, pharmacokinetics, metabolism and toxicity are demonstrated. An expanded model regarding the mode of action of triamterene and its derivatives is also presented.
Product Description
USP
Triamterene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Triamterene, ≥99%
Triamterene, European Pharmacopoeia (EP) Reference Standard