- Review on palladium-containing perovskites: synthesis, physico-chemical properties and applications in catalysis.
Review on palladium-containing perovskites: synthesis, physico-chemical properties and applications in catalysis.
Journal of nanoscience and nanotechnology (2014-04-23)
Abdellatif Essoumhi, Saïd El Kazzouli, Mosto Bousmina
PMID24749470
ABSTRACT
This review reports on the recent advances in the synthesis and physico-chemical properties of palladium-containing perovskites. Initially, the perovskite structure is briefly reviewed, then palladium-containing perovskites synthesis and physico-chemical properties are detailed. The applications of palladium-containing perovskites in catalysis; namely, NO reduction, methane combustion, methanol as well as ethanol oxidation, are briefly highlighted. The involvement and the important contribution of palladium-containing perovskites in cross-coupling reactions, especially Suzuki-Miyaura, Sonogashira, Ulmann and Grignard, are discussed.
Product Description
Palladium, foil, not light tested, 25x25mm, thickness 0.006mm, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.75%
Palladium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95%
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.95%
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 99.95%
Palladium, foil, light tested, 100x100mm, thickness 0.047mm, as rolled, 99.95%
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Palladium, foil, not light tested, 50x50mm, thickness 0.003mm, 99.95%
Palladium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.95%
Palladium, tube, 50mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, as drawn, 99.95%
Palladium, wire reel, 50m, diameter 0.125mm, hard, 99.95%
Palladium, wire reel, 2m, diameter 0.25mm, as drawn, 99.99+%
Palladium, wire reel, 0.5m, diameter 0.02mm, as drawn, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99.95%