MilliporeSigma
  • Fixed-dose combination of phentermine-topiramate for the treatment of obesity.
Fixed-dose combination of phentermine-topiramate for the treatment of obesity.
Expert review of clinical pharmacology (2013-05-10)
Bruno Halpern, André M Faria, Alfredo Halpern
ABSTRACT
The aim of this article is to focus on the fixed-dose combination of phentermine and topiramate, a new antiobesity drug recently approved by the US FDA. The mechanisms of weight loss for each drug in monotherapy is described, followed by the rationale for its use as a combination therapy and a comprehensive review of recently published clinical trials that assessed its efficacy  and safety.
Sigma-Aldrich
Topiramate, ≥98% (HPLC), solid
USP
Topiramate, United States Pharmacopeia (USP) Reference Standard
Supelco
Topiramate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®