MilliporeSigma
Safety of fluconazole in paediatrics: a systematic review.
European journal of clinical pharmacology (2013-01-18)
Oluwaseun Egunsola, Abiodun Adefurin, Apostolos Fakis, Evelyne Jacqz-Aigrain, Imti Choonara, Helen Sammons
ABSTRACT
To determine the safety of fluconazole in neonates and other paediatric age groups by identifying adverse events (AEs) and drug interactions associated with treatment. A search of EMBASE (1950-January 2012), MEDLINE (1946-January 2012), the Cochrane database for systematic reviews and the Cumulative Index to Nursing and Allied Health Literature (1982-2012) for any clinical study about fluconazole use that involved at least one paediatric patient (≤17 years) was performed. Only articles with sufficient quality of safety reporting after patients' exposure to fluconazole were included. We identified 90 articles, reporting on 4,209 patients, which met our inclusion criteria. In total, 794 AEs from 35 studies were recorded, with hepatotoxicity accounting for 378 (47.6 %) of all AEs. When fluconazole was compared with placebo and other antifungals, the relative risk (RR) of hepatotoxicity was not statistically different [RR 1.36, 95 % confidence interval (CI) 0.87-2.14, P = 0.175 and RR 1.43, 95 % CI 0.67-3.03, P = 0.352, respectively]. Complete resolution of hepatoxicity was achieved by 84 % of patients with follow-up available. There was no statistical difference in the risk of gastrointestinal events of fluconazole compared with placebo and other antifungals (RR 0.81, 95 % CI 0.12-5.60, P = 0.831 and RR 1.23, 95 %CI 0.87-1.71, P = 0.235, respectively). There were 41 drug withdrawals, 17 (42 %) of which were due to elevated liver enzymes. Five reports of drug interactions occurred in children. Fluconazole is relatively safe for paediatric patients. Hepatotoxicity and gastrointestinal toxicity are the most common adverse events. It is important to be aware that drug interactions with fluconazole can result in significant toxicity.
Product Description
Supelco
Fluconazole, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fluconazole, United States Pharmacopeia (USP) Reference Standard
Fluconazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Supelco
Fluconazole solution, 2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Fluconazole for peak identification, European Pharmacopoeia (EP) Reference Standard