Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Palladium, foil, 6mm disks, thickness 0.025mm, as rolled, 99.95%
Palladium, foil, 25mm disks, thickness 0.02mm, as rolled, 99.95%
Palladium, foil, 10mm disks, thickness 0.075mm, as rolled, 99.95%
Palladium, foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 99.95%
Palladium, foil, light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, foil, 8mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, wire reel, 5m, diameter 0.025mm, as drawn, 99.9%
Palladium, foil, not light tested, 25x25mm, thickness 0.006mm, 99.95%
Palladium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Palladium, microfoil, disks, 10mm, thinness 0.25μm, specific density 303.4μg/cm2, permanent mylar 3.5μm support, 99.99%
Palladium, wire reel, 2m, diameter 0.25mm, hard, 99.95%
Palladium, microfoil, disks, 25mm, thinness 0.05μm, specific density 63.6μg/cm2, permanent mylar 3.5μm support, 99.99%
Palladium, tube, 50mm, outside diameter 1.0mm, inside diameter 0.8mm, wall thickness 0.1mm, as drawn, 99.95%
Palladium, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95%
Palladium, wire reel, 5m, diameter 0.125mm, hard, 99.95%
Palladium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.95%
Palladium, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.95%