- Biosynthesis of the metalloclusters of molybdenum nitrogenase.
Biosynthesis of the metalloclusters of molybdenum nitrogenase.
Microbiology and molecular biology reviews : MMBR (2011-12-01)
Yilin Hu, Markus W Ribbe
PMID22126998
ABSTRACT
Nitrogenase catalyzes a key step in the global nitrogen cycle, the nucleotide-dependent reduction of atmospheric dinitrogen to bioavailable ammonia. There is a substantial amount of interest in elucidating the biosynthetic mechanisms of the FeMoco and the P-cluster of nitrogenase, because these clusters are not only biologically important but also chemically unprecedented. In this review, we summarize the recent advances in this research area, with an emphasis on our work that aims at providing structural and spectroscopic insights into the assembly of these complex metalloclusters.
Product Description
Molybdenum, foil, 25mm disks, thickness 0.15mm, annealed, 99.9%
Molybdenum, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Molybdenum, foil, light tested, 25x25mm, thickness 0.05mm, annealed, 99.9%
Molybdenum, rod, 500mm, diameter 1.5mm, centerless ground, 99.9%
Molybdenum, tube, 500mm, outside diameter 1.5mm, inside diameter 0.95mm, wall thickness 0.275mm, 99.9%
Molybdenum, rod, 500mm, diameter 2.0mm, centerless ground, 99.9%
Molybdenum, rod, 500mm, diameter 15.0mm, centerless ground, 99.9%
Molybdenum, rod, 50mm, diameter 5.0mm, 99.98%
Molybdenum, rod, 200mm, diameter 5.0mm, centerless ground, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.125mm, annealed, 99.9%
Molybdenum, foil, 5m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Molybdenum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Molybdenum, foil, 100x100mm, thickness 2.0mm, annealed, 99.9%
Molybdenum, wire reel, 200m, diameter 0.2mm, as drawn, 99.95%
Molybdenum, foil, 0.5m coil, thickness 0.0125mm, 99.9%
Molybdenum, wire reel, 5m, diameter 0.015mm, gold plated, 99.95%
Molybdenum, wire reel, 2000m, diameter 0.025mm, hard, 99.95%
Molybdenum, wire reel, 100m, diameter 0.013mm, hard, 99.8+%
Molybdenum, foil, 150x150mm, thickness 0.5mm, annealed, 99.9%
Molybdenum, foil, 25x25mm, thickness 0.25mm, annealed, 99.9%
Molybdenum, foil, 25x25mm, thickness 0.5mm, annealed, 99.9%
Molybdenum, foil, 150x150mm, thickness 0.15mm, annealed, 99.9%