MilliporeSigma
Holmium laser applications of the prostate.
The Urologic clinics of North America (2009-11-28)
Lori B Lerner, Mark D Tyson
ABSTRACT
The high-powered holmium laser is an excellent tool for the surgical treatment of benign prostatic hyperplasia. This article discusses the background of holmium use in the prostate and describes the surgical techniques of holmium laser ablation of the prostate and holmium laser enucleation of the prostate. Operative challenges are reviewed with suggestions as to how to avoid these problems or deal with them when they arise. Surgical outcomes and a thorough literature review are both presented.
Sigma-Aldrich
Holmium, chips, 99.9% trace metals basis
Holmium, rod, 100mm, diameter 6.35mm, cast, 99%
Holmium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Holmium, foil, not light tested, 100x100mm, thickness 0.0125mm, as rolled, 99%
Holmium, rod, 200mm, diameter 3.8mm, 99%
Holmium, rod, 100mm, diameter 3.8mm, 99%
Holmium, rod, 50mm, diameter 3.8mm, 99%
Holmium, foil, not light tested, 50x50mm, thickness 0.0125mm, as rolled, 99%
Holmium, foil, not light tested, 25x25mm, thickness 0.0125mm, as rolled, 99%
Holmium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Holmium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99%
Holmium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Holmium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99%
Holmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Holmium, rod, 50mm, diameter 6.35mm, cast, 99%