MilliporeSigma
  • Hemodynamic and intravascular ultrasound assessment of myocardial bridging: fractional flow reserve paradox with dobutamine versus adenosine.
Hemodynamic and intravascular ultrasound assessment of myocardial bridging: fractional flow reserve paradox with dobutamine versus adenosine.
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions (2009-09-16)
Abdul Hakeem, Mehmet Cilingiroglu, Massoud A Leesar
ABSTRACT
Compared to coronary angiography, both intravascular ultrasound (IVUS) and CT-angiography provide important information with respect to the morphological aspects of myocardial bridging (MB). However, these modalities are limited in defining the hemodynamic and clinical significance of MB. Intracoronary Doppler studies demonstrate a peculiar abnormal Doppler flow profile associated with MB. Fractional flow reserve (FFR) after adenosine infusion has been used to assess the hemodynamic significance of MB, but FFR after adenosine induced hyperemia underestimates the significance of MB. On the other hand, high-dose dobutamine by increasing the contractility of the bridging segment unmasks ischemia induced by MB. This review outlines the role of flow velocity measurement by intracoronary Doppler, FFR, and IVUS for assessment of patients with MB. In addition, we compared FFR measurements after adenosine versus dobutamine infusions for the hemodynamic assessment of MB in two patients.
Product Description
Supelco
Clencyclohexerol hydrochloride, VETRANAL®, analytical standard