MilliporeSigma
  • Early nucleoside reverse transcriptase inhibitors for the treatment of HIV: a brief history of stavudine (D4T) and its comparison with other dideoxynucleosides.
Early nucleoside reverse transcriptase inhibitors for the treatment of HIV: a brief history of stavudine (D4T) and its comparison with other dideoxynucleosides.
Antiviral research (2009-10-27)
John C Martin, Michael J M Hitchcock, Erik De Clercq, William H Prusoff
ABSTRACT
The occasion of this 25th anniversary issue encouraged us to reminisce about the important history of the discovery of the dideoxynucleoside analogues for the treatment of HIV/AIDS and to chronicle our thoughts about a particular exciting and rewarding period of our scientific careers. Following the identification of the anti-HIV activity of zidovudine (AZT), we participated in the urgent quest to discover optimal treatments of HIV infection and AIDS. A number of previously synthesized nucleoside analogues were comparatively evaluated, and stavudine (D4T) emerged as a promising candidate for development. Following clinical evaluation, D4T became a mainstay of the initial antiretroviral combination therapy, prolonging and saving numerous lives. It has only recently been supplanted by better-tolerated treatments. This article forms part of a special issue of Antiviral Research marking the 25th anniversary of antiretroviral drug discovery and development, vol. 85, issue 1, 2010.
Sigma-Aldrich
2′,3′-Didehydro-3′-deoxythymidine, ≥98% (TLC)
Stavudine, European Pharmacopoeia (EP) Reference Standard
Stavudine for system suitability, European Pharmacopoeia (EP) Reference Standard