MilliporeSigma
  • Does acetyl salicylic acid (ASA) have a role in the prevention of venous thromboembolism?
Does acetyl salicylic acid (ASA) have a role in the prevention of venous thromboembolism?
British journal of haematology (2009-05-15)
Ganesan Karthikeyan, John W Eikelboom, Alexander G G Turpie, Jack Hirsh
ABSTRACT
Guidelines differ on whether acetyl salicylic acid (ASA, aspirin) should be used for prophylaxis in patients at high-risk of venous thromboembolism (VTE), principally because of differences in perceptions of its efficacy. ASA is an attractive therapeutic option because it is inexpensive, easy to administer and does not require monitoring. We critically reappraised the evidence from randomized controlled trials for the efficacy of ASA in VTE prevention. ASA is clearly efficacious in preventing VTE compared to placebo or no treatment, but appears to be less efficacious than the low molecular weight heparins in small trials. There is little data for ASA in comparison with unfractionated heparin and warfarin. A large randomized controlled trial is required to clarify the role of ASA compared to contemporary anticoagulant strategies for the prevention of VTE.
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%