MilliporeSigma
Impact of carvedilol on the serum lipid profile.
The Annals of pharmacotherapy (2008-03-28)
Randall P Sharp, Riaz Sirajuddin, Imran M Sharief
ABSTRACT
To examine the evidence regarding the impact of carvedilol on the serum lipid profile. Searches in MEDLINE and International Pharmaceutical Abstracts (1966-December 2007) were conducted. Search terms included carvedilol, cholesterol, lipids, hyperlipidemia, and beta-blockers. Published studies and case reports that evaluated the impact of carvedilol on the lipid profile were reviewed. One study was excluded because it evaluated carvedilol for an off-label use and was a small Phase 2 pilot study that evaluated the results of only 10 patients. Twelve studies were available for review; 6 of these compared carvedilol with beta(1)-selective antagonists. Three studies compared carvedilol with other antihypertensive medications; 2 of those studies evaluated total cholesterol only. Carvedilol alone was evaluated for its effects on lipids in 3 small single-group studies. In 4 of the 12 studies, carvedilol independently improved the lipid profile significantly, while the drug had a nonsignificant, neutral effect on the lipid profile in 3 studies. Furthermore, in 4 of the 12 studies, carvedilol was compared with other antihypertensive medications. In 3 of these 4 studies, the other drugs worsened the lipid profile significantly compared with carvedilol, while carvedilol significantly improved the lipid profile in the other study. Finally, carvedilol had a potentially negative effect on high-density lipoprotein cholesterol in a single-group study, but p values were not reported. It is clear that beta(1)-selective antagonists worsen the lipid profile compared with carvedilol. However, it is unclear whether carvedilol independently makes an improvement or has a neutral effect on the lipid profile. Carvedilol should be an important treatment consideration in patients with heart failure and/or hypertension with dyslipidemia. However, many questions remain regarding this issue.
Sigma-Aldrich
Carvedilol, ≥98% (HPLC), solid
Supelco
Carvedilol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carvedilol, United States Pharmacopeia (USP) Reference Standard
Carvedilol for system suitability, European Pharmacopoeia (EP) Reference Standard
Carvedilol, European Pharmacopoeia (EP) Reference Standard