MilliporeSigma
Carbapenems and monobactams: imipenem, meropenem, and aztreonam.
Mayo Clinic proceedings (1999-04-30)
W C Hellinger, N S Brewer
ABSTRACT
Imipenem and meropenem, members of the carbapenem class of beta-lactam antibiotics, are among the most broadly active antibiotics available for systemic use in humans. They are active against streptococci, methicillin-sensitive staphylococci, Neisseria, Haemophilus, anaerobes, and the common aerobic gram-negative nosocomial pathogens including Pseudomonas. Resistance to imipenem and meropenem may emerge during treatment of P. aeruginosa infections, as has occurred with other beta-lactam agents; Stenotrophomonas maltophilia is typically resistant to both imipenem and meropenem. Like the penicillins, the carbapenems have inhibitory activity against enterococci. In general, the in vitro activity of imipenem against aerobic gram-positive cocci is somewhat greater than that of meropenem, whereas the in vitro activity of meropenem against aerobic gram-negative bacilli is somewhat greater than that of imipenem. Daily dosages may range from 0.5 to 1 g every 6 to 8 hours in patients with normal renal function; the daily dose of meropenem, however, can be safely increased to 6 g. Infusion-related nausea and vomiting, as well as seizures, which have been the main toxic effects of imipenem, occur no more frequently during treatment with meropenem than during treatment with other beta-lactam antibiotics. The carbapenems should be considered for treatment of mixed bacterial infections and aerobic gram-negative bacteria that are not susceptible to other beta-lactam agents. Indiscriminate use of these drugs will promote resistance to them. Aztreonam, the first marketed monobactam, has activity against most aerobic gram-negative bacilli including P. aeruginosa. The drug is not nephrotoxic, is weakly immunogenic, and has not been associated with disorders of coagulation. Aztreonam may be administered intramuscularly or intravenously; the primary route of elimination is urinary excretion. In patients with normal renal function, the recommended dosing interval is every 8 hours. Patients with renal impairment require dosage adjustment. Aztreonam is used primarily as an alternative to aminoglycosides and for the treatment of aerobic gram-negative infections. It is often used in combination therapy for mixed aerobic and anaerobic infections. Approved indications for its use include infections of the urinary tract or lower respiratory tract, intra-abdominal and gynecologic infections, septicemia, and cutaneous infections caused by susceptible organisms. Concurrent initial therapy with other antimicrobial agents is recommended before the causative organism has been determined in patients who are seriously ill or at risk for gram-positive or anaerobic infection.
Product Description
USP
Aztreonam, United States Pharmacopeia (USP) Reference Standard
Supelco
Aztreonam, analytical standard