MilliporeSigma
Synthesis of new hypoxia markers EF1 and [18F]-EF1.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (1999-12-03)
A V Kachur, W R Dolbier, S M Evans, C Y Shiue, G G Shiue, K A Skov, I R Baird, B R James, A R Li, A Roche, C J Koch
ABSTRACT
We report on the preparation of a hypoxia marker 2-(2-nitroimidazol-1[H]-yl)-N-(3-fluoropropyl)acetamide (EF1) and its 18F analog, 2-(2-nitroimidazol-1[H]-yl)-N- (3-[18F]fluoropropyl)acetamide ([18F]-EF1). Two methods for the preparation of 3-fluoropropylamine, the EF1 side chain, are described. [18F]-EF1 was prepared with a radiochemical yield of 2% by nucleophilic substitution of bromine in 2-(2-nitroimidazol-1[H]-yl)-N-(3-bromopropyl)acetamide (EBr1) by carrier-added 18F in DMSO at 120 degrees C. Our results demonstrate the preparation of clinically relevant amounts of [18F]-EF1 for use as a non-invasive hypoxia marker with detection using positron emission tomography (PET).
Product Description
Supelco
AmberChrom 1X4 chloride form, 200-400 mesh, Anion Exchange Resin
Supelco
AmberChrom 1X4 chloride form, chloride form, 100-200 mesh
Supelco
AmberChrom 1X4 chloride form, 20-50 mesh, Anion Exchange Resin