MilliporeSigma
  • The TNFSF15 gene single nucleotide polymorphism rs7848647 is associated with surgical diverticulitis.
The TNFSF15 gene single nucleotide polymorphism rs7848647 is associated with surgical diverticulitis.
Annals of surgery (2014-05-13)
Tara M Connelly, Arthur S Berg, John P Hegarty, Sue Deiling, David Brinton, Lisa S Poritz, Walter A Koltun
ABSTRACT
To determine if single nuclear polymorphisms (SNPs) in the TFNSF15 gene play a role in patients requiring surgery for diverticulitis. A role for a genetic predisposition in diverticulitis is suggested by its association with hereditary connective tissue disorders, youthful onset in some patients, and the observation of families with multiple affected individuals. The TNFSF15 gene has been associated with other inflammatory diseases affecting the colon such as medically refractory ulcerative colitis (UC), aggressive Crohn's disease (CD), and pouchitis after restorative proctocolectomy. In the discovery phase of this study, 21 sporadic surgical diverticulitis (SD) patients (9 female, mean age = 52 ± 5) and 5 individuals from a single family with surgically managed diverticulitis [familial diverticulitis (FD), 4 female, mean age = 51.1 ± 7] were studied. SD patients were age and sex matched with 3 separate groups of healthy, CD and UC control patients. All patients were genotyped for 5 known TNFSF15-associated SNPs. The SNP discovered to be associated with diverticulitis (rs7848647) was then confirmed in a separate test group composed of 34 additional patients (20 female, mean age 57.7 ± 2) who also underwent surgical treatment for diverticulitis. These patients were age matched to a new control cohort of patients having no history of diverticulitis (26 female). Patients were genotyped using a TaqMan assay. In the discovery phase, logistical regression on matched subjects was performed to determine an association of TNFSF SNP with diverticulitis versus the control groups. In the test phase, significance for the rs7848647 SNP was assessed by the Fischer's exact test. In the discovery phase, the TNFSF15 SNP rs7848647 was significantly associated with SD (p = 0.0003) versus all control groups studied. The risk allele for this SNP (G substituted for A) was found in all SD patients. The homozygous GG allele was found in 62% (13/21) of SD patients versus only 5% (1/21) of healthy controls (p = 0.001) and 24% (10/42) of all UC + CD controls (p = 0.002). All 5 members of the FD cohort were homozygous for the at-risk "G" allele. In the test group, the homozygous GG genotype was found in 56% of SD patients compared with 17% of healthy controls (p = 0.006). Risk of SD seemed to increase with number of the G alleles with 8% of SD patients having AA homozygosity, 35% of SD patients having AG heterozygosity, and 56% of SD patients having GG homozygosity. The SNP rs7848647 associated with the TNFSF15 gene is associated with surgical diverticulitis. This finding suggests a fundamental role for TNFSF15, a T-cell receptor gene involved in T-cell maturation, in the pathophysiology of diverticulitis requiring surgery. This SNP may be a marker of diverticular disease severity that might assist in surgical decision making.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from herring testes, Type XIV
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC19, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid from human placenta, buffered aqueous solution, sexed, female
Sigma-Aldrich
Plasmid DNA from E. coli RRI, pUC18, buffered aqueous solution
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from salmon testes, For hybridization