MilliporeSigma
  • Formulation and optimization of raloxifene-loaded solid lipid nanoparticles to enhance oral bioavailability.
Formulation and optimization of raloxifene-loaded solid lipid nanoparticles to enhance oral bioavailability.
Journal of nanoscience and nanotechnology (2014-04-25)
Tuan Hiep Tran, Thiruganesh Ramasamy, Hyuk Jun Cho, Yong Il Kim, Bijay Kumar Poudel, Han-Gon Choi, Chul Soon Yong, Jong Oh Kim
ABSTRACT
The main aim of this study was to improve the oral bioavailability of raloxifene (RXF), a selective estrogen receptor modulator, by incorporation into solid lipid nanoparticles (SLN). RXF-loaded SLN was prepared by homogenization-sonication technique and characterized through physicochemical, pharmacokinetic, and cytotoxicity studies. The optimized SLN formulation exhibited a spherical shape with average size around 140 nm, easing its transport across the lymphatic system. Augmentation in the profiles of C(max) (308%) and AUC (270%) indicated a significant enhancement in the rate and extent of bioavailability by SLN formulations compared to free drug. In vitro cytotoxicity study performed in NIH-3T3 cells revealed that RXF-SLN was cytocompatible, and SLN remained unchanged during the freeze-drying process. Furthermore, the optimized formulation was quite stable at room temperature for more than two months, exemplifying its superior performance. In conclusion, SLN provides a promising platform for the pronounced enhancement of RXF bioavailability.
Product Description
Raloxifene hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Raloxifene hydrochloride, solid
USP
Raloxifene hydrochloride, United States Pharmacopeia (USP) Reference Standard
Raloxifene hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard