MilliporeSigma
  • Leptin and IL-8: two novel cytokines screened out in childhood lead exposure.
Leptin and IL-8: two novel cytokines screened out in childhood lead exposure.
Toxicology letters (2014-04-09)
Yi Yang, Xuezhong Zhang, Yawen Fu, Hui Yang
ABSTRACT
Lead is a toxic heavy metal with many recognized adverse health side effects. The central nervous system is the main target of lead toxicity. Although many studies on lead toxicity were conducted, the mechanism of lead toxicity remains uncertain. One possible attribution is the immature blood-brain barrier that causes lead exposure in children. Few studies have investigated the cytokine changes caused by this exposure. Novel cytokines were detected by RayBio(®) Human Cytokine Antibody Array and validated by enzyme-linked immunosorbent assay. Several children were admitted to West China Second University Hospital, after a serious lead pollution event in longchang, Sichuan, China. A total of 4 children with elevated blood lead levels (BLLs) and 4 children with low BLLs were randomly chosen in the discovery set, and 40 children with elevated BLLs and 40 children with low BLLs were included in the validation set. Leptin and interleukin-8 (IL-8) were identified to be significantly different between children with elevated and low BLLs via RayBio(®) Human Cytokine Antibody Array. In the validation set, IL-8 was higher in children with elevated BLLs [median(P25-P75), 117.69(52.31-233.63) pg/mL] than in children with low BLLs [median(P25-P75): 17.70(10.75-26.52) pg/mL] (p=0.000). Leptin was lower in children with elevated BLLs [median(P25-P75): 1658.23(1421.86-2606.55) pg/mL] than in children with low BLLs [median(P25-P75): 4168.68(3246.32-4744.94) pg/mL] (p=0.000). In children with low BLLs, leptin was higher in children with BLLs<3 μg/dL (N=7) [median(P25-P75): 7220.86(4265.72-7555.15) pg/mL] than in children with BLL ≥ 3 μg/dL (N=33) [median(P25-P75): 4103.86(3163.40-4678.34) pg/mL] (p=0.026); IL-8 was significantly different in children with BLL<4 μg/dL (N=13) [median(P25-P75): 12.49(8.25-14.86) pg/mL] than in children with BLL≥4 μg/dL (N=27) [median(P25-P75): 21.98(13.64-33.50) pg/mL] (p=0.013). The results defined specific changes in cytokine expressions to lead exposure, which can be used to explore the mechanism of lead toxicity and monitor lead exposure.
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Lead, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 50x50mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 150x150mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 6mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 1m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 5m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 1m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, light tested, 100x100mm, thickness 0.025mm, 99.99+%
Lead, foil, 100x100mm, thickness 1.5mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.007mm, 99.99+%
Lead, foil, 6mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 1m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 15mm disks, thickness 0.009mm, 99.99+%
Lead, foil, light tested, 100x100mm, thickness 0.04mm, 99.99+%
Lead, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 6mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.55mm, as rolled, 100%
Lead, foil, 10mm disks, thickness 0.008mm, 99.99+%
Lead, foil, 500x500mm, thickness 3.0mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.04mm, 99.99+%