MilliporeSigma
  • Lead and other elements in house dust of Japanese residences--source of lead and health risks due to metal exposure.
Lead and other elements in house dust of Japanese residences--source of lead and health risks due to metal exposure.
Environmental pollution (Barking, Essex : 1987) (2014-04-01)
Jun Yoshinaga, Kumiko Yamasaki, Ayumi Yonemura, Yuri Ishibashi, Takaya Kaido, Kodai Mizuno, Mai Takagi, Atsushi Tanaka
ABSTRACT
The levels of 25 elements in house dust collected from 100 general Japanese residences were measured. Factor analysis was applied on the multi-element data to explore source of Pb (median concentration 49.1 mg/kg) in house dust. Six factors were extracted and Pb was found to have great loading on the fifth factor with Sb and Sn, suggesting solder (Sn), and plastic and metals (Sb) may be the sources of Pb in the house dust of Japanese residences. No significant loading was found on soil-related factors indicating non-significant contribution of Pb in track-in soil. Seven heavy metals (Cd, Cu, Mo, Pb, Sb, Sn, and Zn) were found in house dust at >10 times more condensed than crustal abundance. Health risk of these elements to children via the ingestion of house dust was estimated based on the comparison with tolerable daily intake and found to be non-significant for most of the elements.
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Lead, foil, 2m coil, thickness 0.15mm, as rolled, 99.95%
Lead, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.999%
Lead, foil, not light tested, 25x25mm, thickness 0.02mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.025mm, 99.99+%
Lead, wire reel, 1m, diameter 0.5mm, as drawn, 99.999%
Lead, foil, 25mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.5mm, as rolled, 99.95%
Lead, foil, not light tested, 100x100mm, thickness 0.008mm, 99.99+%
Lead, foil, 50x50mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.05mm, 99.99+%
Lead, foil, 6mm disks, thickness 0.03mm, 99.99+%
Lead, foil, 1m coil, thickness 1.0mm, as rolled, 99.95%
Lead, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, not light tested, 100x100mm, thickness 0.01mm, 99.99+%
Lead, rod, 1000mm, diameter 6.35mm, 99.95%
Lead, foil, not light tested, 50x50mm, thickness 0.007mm, 99.99+%
Lead, rod, 500mm, diameter 19.0mm, 99.95%
Lead, foil, light tested, 25x25mm, thickness 0.007mm, 99.99+%
Lead, rod, 100mm, diameter 12.7mm, 99.95%
Lead, rod, 500mm, diameter 3.2mm, 99.95%
Lead, foil, not light tested, 300x300mm, thickness 0.016mm, as rolled, 96% (Sn 1.5%, Sb 2.5%)