MilliporeSigma
  • Risk of myeloproliferative disease and chronic myeloid leukaemia following exposure to low-level benzene in a nested case-control study of petroleum workers.
Risk of myeloproliferative disease and chronic myeloid leukaemia following exposure to low-level benzene in a nested case-control study of petroleum workers.
Occupational and environmental medicine (2014-02-18)
Deborah C Glass, A Robert Schnatter, Gong Tang, Richard D Irons, Lesley Rushton
ABSTRACT
Benzene exposure has been associated with increased risk of leukaemia and myelodysplastic syndrome. Existing studies are sparse for other lymphohaematopoietic cancer subtypes, such as myeloproliferative disease (MPD) and the related chronic myeloid leukaemia (CML). We pooled data from three petroleum worker nested case-control studies to address this gap. To our knowledge, this is the first study to systematically examine the relationship between MPD and quantitative benzene exposure. There were 28 cases and 122 matched controls for CML and 30 MPD cases with 124 matched controls. Two haematopathologists identified each case and provided a diagnosis certainty score. Blinded data-driven assessments estimated benzene exposure for each job held by study participants. Statistical analyses included conditional logistic regression and penalised smoothing splines. Benzene exposures were low, and mean average exposure intensity for CML cases was 0.3 ppm and for MPD cases 0.17 ppm. Categorical analyses showed no increased risk of CML or MPD with benzene exposure. There was no significantly increased risk identified for more highly exposed terminal workers. Some association was seen in spline analyses between increased risk of MPD and benzene exposure experienced in the 2-20 years before diagnosis and with peak exposures considered with cumulative exposure as a continuous variable. No convincing association was identified between MPD or CML and low exposure to benzene. The greater risk for exposures experienced in the 20 years before diagnosis needs investigating in more powerful studies with a wider range of exposure to benzene, and the biological plausibility further examined from a mechanistic viewpoint.
Product Description
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Benzene, SAJ first grade, ≥99.0%
Sigma-Aldrich
Benzene, JIS 300, ≥99.5%, for residue analysis
Sigma-Aldrich
Benzene, JIS special grade, ≥99.5%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol