MilliporeSigma
  • Metabolic perturbation of an essential pathway: evaluation of a glycine precursor of coenzyme A.
Metabolic perturbation of an essential pathway: evaluation of a glycine precursor of coenzyme A.
Journal of the American Chemical Society (2013-04-05)
Michael Rothmann, MinJin Kang, Reymundo Villa, Ioanna Ntai, James J La Clair, Neil L Kelleher, Eli Chapman, Michael D Burkart
ABSTRACT
Pantetheine and its corresponding disulfide pantethine play a key role in metabolism as building blocks of coenzyme A (CoA), an essential cofactor utilized in ~4% of primary metabolism and central to fatty acid, polyketide, and nonribosomal peptide synthases. Using a combination of recombinant engineering and chemical synthesis, we show that the disulfide of N-pantoylglycyl-2-aminoethanethiol (GlyPan), with one fewer carbon than pantetheine, can rescue a mutant E. coli strain MG1655ΔpanC lacking a functional pantothenate synthetase. Using mass spectrometry, we show that the GlyPan variant is accepted by the downstream CoA biosynthetic machinery, ultimately being incorporated into essential acyl carrier proteins. These findings point to further flexibility in CoA-dependent pathways and offer the opportunity to incorporate orthogonal analogues.
Sigma-Aldrich
(R)-Pantetheine, ≥95.0% (HPLC)
Sigma-Aldrich
(R)-Pantoic acid sodium salt, ≥95% (GC)