MilliporeSigma
Synthesis of (+/-)-vibralactone.
Organic letters (2008-03-04)
Quan Zhou, Barry B Snider
ABSTRACT
Reductive alkylation of methyl 2-methoxybenzoate with prenyl bromide and hydrolysis afforded methyl 6-oxo-1-prenyl-2-cyclohexenecarboxylate. Reduction of the ketone, hydrolysis, iodolactonization, ozonolysis, and intramolecular aldol reaction provided a spiro lactone cyclopentenal. Retro-iodolactonization with activated Zn, formation of the beta-lactone, and reduction of the aldehyde completed an efficient first synthesis of (+/-)-vibralactone. No protecting groups were used except for the novel use of an iodolactone to protect both the prenyl double bond and carboxylic acid.
Sigma-Aldrich
Methyl 2-methoxybenzoate, 99%