MilliporeSigma
  • Highly efficient synthesis of (R)-3-quinuclidinol in a space-time yield of 916 g L(-1) d(-1) using a new bacterial reductase ArQR.
Highly efficient synthesis of (R)-3-quinuclidinol in a space-time yield of 916 g L(-1) d(-1) using a new bacterial reductase ArQR.
Organic letters (2013-09-21)
Wen-Xia Zhang, Guo-Chao Xu, Lei Huang, Jiang Pan, Hui-Lei Yu, Jian-He Xu
ABSTRACT
A new keto reductase (ArQR), identified from Agrobacterium radiobacter ECU2556, can efficiently reduce 3-quinuclidinone in excellent enantioselectivity and high space-time yield for the synthesis of (R)-3-quinuclidinol, a chiral building block of many antimuscarinic agents. This is the first time that a high yield of (R)-3-quinuclidinol up to 916 g L(-1) d(-1) using a bioreduction approach is reported.
Sigma-Aldrich
3-Quinuclidinol, 99%