MilliporeSigma
  • Conventional and fast gas chromatography analysis of biodiesel blends using an ionic liquid stationary phase.
Conventional and fast gas chromatography analysis of biodiesel blends using an ionic liquid stationary phase.
Journal of chromatography. A (2009-11-17)
Carla Ragonese, Peter Quinto Tranchida, Danilo Sciarrone, Luigi Mondello
ABSTRACT
The present research is focused on the GC-FID determination of fatty acid methyl esters (FAMEs) in diesel blends, by means of an ionic liquid stationary phase, characterized by a dicationic 1,9-di(3-vinyl-imidazolium)nonane bis(trifluoromethyl)sulfonylimidate structure (SLB-IL100). The high polarity of the ionic liquid stationary phase allowed the separation of the FAMEs, from the less-retained hydrocarbons, thus avoiding the requirement of a hydrocarbon LC pre-separation. The results derived from the analyses of a soybean FAMEs B20 sample, carried out on an SLB-IL100 conventional column (30 m x 0.25 mm i.d. x 0.20 mm d(f)), were compared with those attained on a polyethylene glycol column, of equivalent dimensions. Conventional and fast GC methods, for the analysis of FAMEs in diesel blends, were developed on an SLB-IL100 30 m x 0.25 mm i.d. x 0.20 microm d(f) and on an SLB-IL100 12 m x 0.10 mm i.d. x 0.08 microm d(f) column, respectively. The optimized IL methods were subjected to validation: retention time and peak area intra-day precision (n=5) were good, with CV % values lower than 0.08% and 4.9%, respectively. With regards to the quantitation of FAMEs in biodiesel blends, a five points calibration curve was constructed, using C(17:0) as internal standard.
Product Description
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub