MilliporeSigma
  • Quantifying lipid diffusion by fluorescence correlation spectroscopy: a critical treatise.
Quantifying lipid diffusion by fluorescence correlation spectroscopy: a critical treatise.
Langmuir : the ACS journal of surfaces and colloids (2012-08-16)
Fabian Heinemann, Viktoria Betaneli, Franziska A Thomas, Petra Schwille
ABSTRACT
Fluorescence correlation spectroscopy (FCS) measurements are widely used for determination of diffusion coefficients of lipids and proteins in biological membranes. In recent years, several variants of FCS have been introduced. However, a comprehensive comparison of these methods on identical systems has so far been lacking. In addition, there exist no consistent values of already determined diffusion coefficients for well-known or widely used membrane systems. This study aims to contribute to a better comparability of FCS experiments on membranes by determining the absolute diffusion coefficient of the fluorescent lipid analog 1,1'-dioctadecyl-3,3,3',3'-tetramethylindodicarbocyanine (DiD) in giant unilamellar vesicles (GUVs) made of dioleoylphosphatidylcholine (DOPC), which can in future studies be used as a reference value. For this purpose, five FCS variants, employing different calibration methods, were compared. Potential error sources for each particular FCS method and strategies to avoid them are discussed. The obtained absolute diffusion coefficients for DiD in DOPC were in good agreement for all investigated FCS variants. An average diffusion coefficient of D = 10.0 ± 0.4 μm(2) s(-1) at 23.5 ± 1.5 °C was obtained. The independent confirmation with different methods indicates that this value can be safely used for calibration purposes. Moreover, the comparability of the methods also in the case of slow diffusion was verified by measuring diffusion coefficients of DiD in GUVs consisting of DOPC and cholesterol.
Sigma-Aldrich
Atto 655-Biotin, BioReagent, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Atto 655, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Atto 655-maleimide, BioReagent, suitable for fluorescence, ≥90% (coupling rate)
Sigma-Aldrich
Atto 655 NHS ester, BioReagent, suitable for fluorescence