MilliporeSigma
  • Retention of iodinated contrast material within renal cysts in a patient with autosomal dominant polycystic kidney disease.
Retention of iodinated contrast material within renal cysts in a patient with autosomal dominant polycystic kidney disease.
The British journal of radiology (2012-03-07)
S J Park
ABSTRACT
Hyperdense renal cysts, a common condition in autosomal dominant polycystic kidney disease, may be induced by haemorrhage into the cysts. However, hyperdense renal cysts resulting from retention of contrast material after intravenous injection is extremely uncommon because the intravenous administration of contrast material does not induce an increase in the attenuation of renal cysts. We report a case of retention of iodinated contrast material within renal cysts in a patient with autosomal dominant polycystic kidney disease.
Product Description
Ioxaglic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ioxaglic acid, United States Pharmacopeia (USP) Reference Standard