MilliporeSigma
  • Effects of a constant rate infusion of detomidine on cardiovascular function, isoflurane requirements and recovery quality in horses.
Effects of a constant rate infusion of detomidine on cardiovascular function, isoflurane requirements and recovery quality in horses.
Veterinary anaesthesia and analgesia (2011-10-13)
Stijn Schauvliege, Miguel Gozalo Marcilla, Kirsten Verryken, Luc Duchateau, Lindsey Devisscher, Frank Gasthuys
ABSTRACT
To examine the influence of a detomidine constant rate infusion (CRI) on cardiovascular function, isoflurane requirements and recovery quality in horses undergoing elective surgery. Prospective, randomized, blinded, clinical trial. Twenty adult healthy horses. After sedation (detomidine, 10 μg kg(-1) intravenously [IV]) and induction of anaesthesia (midazolam 0.06 mg kg(-1) , ketamine 2.2 mg kg(-1) IV), anaesthesia was maintained with isoflurane in oxygen/air (inspiratory oxygen fraction 55%). When indicated, the lungs were mechanically ventilated. Dobutamine was administered when MAP<70 mmHg. The horses were randomly allocated to one of two groups and throughout anaesthesia, received either a detomidine (5 μg kg(-1)  hour(-1) ) (D) or saline (S) CRI, with the anaesthetist unaware of the treatment. Monitoring included end-tidal isoflurane concentration, arterial pH, PaCO(2) , PaO(2) , dobutamine administration rate, heart rate (HR), arterial pressure, cardiac index (CI), systemic vascular resistance (SVR), stroke index and oxygen delivery index (ḊO(2) I). For recovery from anaesthesia, all horses received 2.5 μg kg(-1) detomidine IV. Recovery quality and duration were recorded in each horse. For statistical analysis, anova, Pearson chi-square and Wilcoxon rank sum tests were used as relevant. Heart rate (p=0.0176) and ḊO(2) I (p= 0.0084) were lower and SVR higher (p=0.0126) in group D, compared to group S. Heart rate (p=0.0011) and pH (p=0.0187) increased over time. Significant differences in isoflurane requirements were not detected. Recovery quality and duration were comparable between treatments. A detomidine CRI produced cardiovascular effects typical for α(2) -agonists, without affecting isoflurane requirements, recovery duration or recovery quality.
Product Description
Detomidine hydrochloride, European Pharmacopoeia (EP) Reference Standard