MilliporeSigma
  • Recent advances in semisynthesis, biosynthesis, biological activities, mode of action, and structure-activity relationship of podophyllotoxins: an update (2008-2010).
Recent advances in semisynthesis, biosynthesis, biological activities, mode of action, and structure-activity relationship of podophyllotoxins: an update (2008-2010).
Mini reviews in medicinal chemistry (2011-07-26)
M Lv, H Xu
ABSTRACT
Podophyllotoxin, one of the well-known naturally occurring aryltetralin lignans, has been used as the lead-compound for the preparation of potent anticancer agents, such as etoposide, teniposide, and etopophos. In our previous review, we described the advances of podophyllotoxin derivatives from 2003 and 2007. In recent years, an increased number of interesting research work has been carried out on the podophyllotoxins. As a continuation, the present review summarizes and highlights the update advances of podophyllotoxin derivatives from 2008 and 2010 in regard to semisynthesis, biosynthesis, biological activities, mode of action and structure-biological activity relationship.
Sigma-Aldrich
Teniposide, ≥97% (HPLC)