MilliporeSigma
  • Imaging of adrenal incidentalomas with PET using (11)C-metomidate and (18)F-FDG.
Imaging of adrenal incidentalomas with PET using (11)C-metomidate and (18)F-FDG.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2004-06-08)
Heikki Minn, Anna Salonen, Johan Friberg, Anne Roivainen, Tapio Viljanen, Jaakko Långsjö, Jorma Salmi, Matti Välimäki, Kjell Någren, Pirjo Nuutila
ABSTRACT
Our aim was to evaluate the use of PET with (11)C-metomidate and (18)F-FDG for the diagnosis of adrenal incidentalomas. Twenty-one patients underwent hormonal screening before dynamic imaging of the upper abdomen with (11)C-metomidate, and for 19 of these 21 patients, static (18)F-FDG imaging followed. Uptake of (11)C-metomidate and (18)F-FDG in incidentalomas was quantified and correlated with the hormonal work-up and the mass size on CT (median, 2.5 cm; range, 2-10 cm). The final diagnoses were hormonally active adenoma (n = 7), nonsecretory adenoma (n = 5), adrenocortical carcinoma (n = 1), pheochromocytoma (n = 2), benign noncortical tumor (n = 2), normal adrenal (n = 1), and malignant noncortical tumor (n = 3). Diagnosis was established at surgery (n = 9), percutaneous biopsy (n = 4), or follow-up (n = 8). The highest uptake of (11)C-metomidate, expressed as standardized uptake value (SUV), was found in adrenocortical carcinoma (SUV = 28.0), followed by active adenomas (median SUV = 12.7), nonsecretory adenomas (median SUV = 12.2), and noncortical tumors (median SUV = 5.7). Patients with adenomas had significantly higher tumor-to-normal-adrenal (11)C-metomidate SUV ratios than did patients with noncortical tumors. (18)F-FDG detected 2 of 3 noncortical malignancies but failed to detect adrenal metastases from renal cell carcinoma. All inactive and most active adenomas were difficult to detect with (18)F-FDG against background activity, whereas both pheochromocytomas and adrenocortical carcinoma showed slightly increased uptake of (18)F-FDG. There was no correlation between uptake of (11)C-metomidate or (18)F-FDG and mass size. (11)C-Metomidate is a promising PET tracer to identify incidentalomas of adrenocortical origin. (18)F-FDG should be reserved for patients with a moderate to high likelihood of neoplastic disease.
Product Description
Etomidate impurity B, European Pharmacopoeia (EP) Reference Standard