MilliporeSigma
The court as a catalyst in the treatment process.
American journal of psychotherapy (1988-04-01)
G A Awad, C Perillo
ABSTRACT
The role of the Court in the treatment process varies. In some cases it can work as a catalyst. We present the treatment of a family with a mother whose involving psychosis kept her daughter from attending school, and show how the power of the Court can be useful.
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥85% (PAGE), lyophilized powder
Sigma-Aldrich
β-Lactoglobulin B from bovine milk, ≥90% (PAGE)
Sigma-Aldrich
β-Lactoglobulin A from bovine milk, ≥90% (PAGE)
Sigma-Aldrich
β-Lactoglobulin A from bovine milk, Isoelectric focusing marker, pI 5.1