MilliporeSigma
  • Development of a quantitative PCR (qPCR) for Giardia and analysis of the prevalence, cyst shedding and genotypes of Giardia present in sheep across four states in Australia.
Development of a quantitative PCR (qPCR) for Giardia and analysis of the prevalence, cyst shedding and genotypes of Giardia present in sheep across four states in Australia.
Experimental parasitology (2013-12-21)
Rongchang Yang, Caroline Jacobson, Graham Gardner, Ian Carmichael, Angus J D Campbell, Una Ryan
ABSTRACT
A novel quantitative PCR (qPCR) for Giardia at the glutamate dehydrogenase (gdh) locus was developed and validated. The qPCR was used to screen a total of 3412 lamb faecal samples collected from approximately 1189 lambs at three sampling periods (weaning, post-weaning and pre-slaughter) from eight farms across South Australia (SA), New South Wales (NSW), Victoria (Vic) and Western Australia (WA). The overall prevalence was 20.2% (95% CI 18.9-21.6) and of the 690 positives, 473 were successfully typed. In general, the prevalence of Giardia varied widely across the different farms with the highest prevalence in one WA farm (42.1%) at pre-slaughter sampling and the lowest prevalence in one Victorian farm (7.2%) at weaning. The range of cyst shedding at weaning, post-weaning and pre-slaughter overall across all states was 63-1.3×10(9) cysts g(-1) (median=1.7×10(4)), 63-1.1×10(9) cysts g(-1) (median=9.6×10(3)), 63-4.7×10(9) cysts g(-1) (median=8.1×10(4)) respectively. Assemblage specific primers at the triose phosphate isomerase (tpi) locus identified assemblage A in 22.4% (106/473) of positive samples typed, assemblage E in 75.9% (359/473) and mixed A and E assemblages in 1.7% (8/473) of samples. A subset of representative samples from the 8 farms (n=32) were typed at both the gdh and beta-giardin loci and confirmed these results and identified sub-assemblage AII in 16 representative assemblage A isolates across the 8 farms. This demonstrates a prevalence of Giardia previously not recognised in Australian sheep, highlighting a need for further research to quantify the production impacts of this protozoan parasite.
Sigma-Aldrich
Triosephosphate Isomerase from baker′s yeast (S. cerevisiae), Type I, ammonium sulfate suspension, ~10,000 units/mg protein
Sigma-Aldrich
Triosephosphate Isomerase from rabbit muscle, Type X, lyophilized powder, ≥3,500 units/mg protein
Sigma-Aldrich
Triosephosphate Isomerase from rabbit muscle, Type III-S, ammonium sulfate suspension, ≥4,000 units/mg protein