MilliporeSigma
  • Parallel measurement of dynamic changes in translation rates in single cells.
Parallel measurement of dynamic changes in translation rates in single cells.
Nature methods (2013-11-12)
Kyuho Han, Ariel Jaimovich, Gautam Dey, Davide Ruggero, Oded Meyuhas, Nahum Sonenberg, Tobias Meyer
ABSTRACT
Protein concentrations are often regulated by dynamic changes in translation rates. Nevertheless, it has been challenging to directly monitor changes in translation in living cells. We have developed a reporter system to measure real-time changes of translation rates in human or mouse individual cells by conjugating translation regulatory motifs to sequences encoding a nuclear targeted fluorescent protein and a controllable destabilization domain. Application of the method showed that individual cells undergo marked fluctuations in the translation rate of mRNAs whose 5' terminal oligopyrimidine (5' TOP) motif regulates the synthesis of ribosomal proteins. Furthermore, we show that small reductions in amino acid levels signal through different mTOR-dependent pathways to control TOP mRNA translation, whereas larger reductions in amino acid levels control translation through eIF2A. Our study demonstrates that dynamic measurements of single-cell activities of translation regulatory motifs can be used to identify and investigate fundamental principles of translation.
Product Description
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Trimethoprim for system suitability, European Pharmacopoeia (EP) Reference Standard