MilliporeSigma
  • Screening, monitoring, and treatment of stage 1 to 3 chronic kidney disease: A clinical practice guideline from the American College of Physicians.
Screening, monitoring, and treatment of stage 1 to 3 chronic kidney disease: A clinical practice guideline from the American College of Physicians.
Annals of internal medicine (2013-10-23)
Amir Qaseem, Robert H Hopkins, Donna E Sweet, Melissa Starkey, Paul Shekelle
ABSTRACT
The American College of Physicians (ACP) developed this guideline to present the evidence and provide clinical recommendations on the screening, monitoring, and treatment of adults with stage 1 to 3 chronic kidney disease. This guideline is based on a systematic evidence review evaluating the published literature on this topic from 1985 through November 2011 that was identified by using MEDLINE and the Cochrane Database of Systematic Reviews. Searches were limited to English-language publications. The clinical outcomes evaluated for this guideline included all-cause mortality, cardiovascular mortality, myocardial infarction, stroke, chronic heart failure, composite vascular outcomes, composite renal outcomes, end-stage renal disease, quality of life, physical function, and activities of daily living. This guideline grades the evidence and recommendations by using ACP's clinical practice guidelines grading system. ACP recommends against screening for chronic kidney disease in asymptomatic adults without risk factors for chronic kidney disease. (Grade: weak recommendation, low-quality evidence) ACP recommends against testing for proteinuria in adults with or without diabetes who are currently taking an angiotensin-converting enzyme inhibitor or an angiotensin II-receptor blocker. (Grade: weak recommendation, low-quality evidence) ACP recommends that clinicians select pharmacologic therapy that includes either an angiotensin-converting enzyme inhibitor (moderate-quality evidence) or an angiotensin II-receptor blocker (high-quality evidence) in patients with hypertension and stage 1 to 3 chronic kidney disease. (Grade: strong recommendation) ACP recommends that clinicians choose statin therapy to manage elevated low-density lipoprotein in patients with stage 1 to 3 chronic kidney disease. (Grade: strong recommendation, moderate-quality evidence).
Sigma-Aldrich
Gemfibrozil
Supelco
Gemfibrozil, Pharmaceutical Secondary Standard; Certified Reference Material
Gemfibrozil, European Pharmacopoeia (EP) Reference Standard
Gemfibrozil for system suitability, European Pharmacopoeia (EP) Reference Standard