MilliporeSigma
  • Renal injury and Nrf2 modulation in mouse kidney following chronic exposure to TiO₂ nanoparticles.
Renal injury and Nrf2 modulation in mouse kidney following chronic exposure to TiO₂ nanoparticles.
Journal of agricultural and food chemistry (2013-08-24)
Suxin Gui, Bingyan Li, Xiaoyang Zhao, Lei Sheng, Jie Hong, Xiaohong Yu, Xuezi Sang, Qingqing Sun, Yuguan Ze, Ling Wang, Fashui Hong
ABSTRACT
TiO₂ nanoparticles (NPs) are used in the food industry but have potential toxic effects in humans and animals. TiO₂ NPs impair renal function and cause oxidative stress and renal inflammation in mice, associated with inhibition of nuclear factor erythroid-2-related factor 2 (Nrf2), which regulates genes encoding many antioxidants and detoxifying enzymes. This study determined whether TiO₂ NPs activated the Nrf2 signaling pathway. Mice exhibited accumulation of reactive oxygen species and peroxidation of lipid, protein, and DNA in the kidney, coupled with renal dysfunction, glutathione depletion, inflammatory cell infiltration, fatty degeneration, and apoptosis. These were associated with increased expression of NOX4, cyclooxygenase-2, and nuclear factor-κB. Oxidative stress and inflammation were accompanied by decreased expression of Nrf2 and down-regulation of its target gene products including heme oxygenase 1, glutamate-cysteine ligase catalytic subunit, and glutathione S-transferase. Chronic TiO₂ NP exposure is associated with suppression of Nrf2, which contributes to the pathogenesis of oxidative stress and inflammation.
Sigma-Aldrich
Titanium(IV) oxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5%
Sigma-Aldrich
Titanium(IV) oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Titanium(IV) oxide, rutile, 99.995% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis