MilliporeSigma
  • Photochemical activity of a key donor-acceptor complex can drive stereoselective catalytic α-alkylation of aldehydes.
Photochemical activity of a key donor-acceptor complex can drive stereoselective catalytic α-alkylation of aldehydes.
Nature chemistry (2013-08-24)
Elena Arceo, Igor D Jurberg, Ana Alvarez-Fernández, Paolo Melchiorre
ABSTRACT
Asymmetric catalytic variants of sunlight-driven photochemical processes hold extraordinary potential for the sustainable preparation of chiral molecules. However, the involvement of short-lived electronically excited states inherent to any photochemical reaction makes it challenging for a chiral catalyst to dictate the stereochemistry of the products. Here, we report that readily available chiral organic catalysts, with well-known utility in thermal asymmetric processes, can also confer a high level of stereocontrol in synthetically relevant intermolecular carbon-carbon bond-forming reactions driven by visible light. A unique mechanism of catalysis is proposed, wherein the catalyst is involved actively in both the photochemical activation of the substrates (by inducing the transient formation of chiral electron donor-acceptor complexes) and the stereoselectivity-defining event. We use this approach to enable transformations that are extremely difficult under thermal conditions, such as the asymmetric α-alkylation of aldehydes with alkyl halides, the formation of all-carbon quaternary stereocentres and the control of remote stereochemistry.
Sigma-Aldrich
2,6-Dimethylpyridine, ≥99%
Sigma-Aldrich
2,6-Lutidine, ReagentPlus®, 98%
Supelco
2,6-Lutidine, analytical standard
Sigma-Aldrich
2,6-Lutidine