MilliporeSigma
  • Fluorescein analogue xanthene-9-carboxylic acid: a transition-metal-free CO releasing molecule activated by green light.
Fluorescein analogue xanthene-9-carboxylic acid: a transition-metal-free CO releasing molecule activated by green light.
Organic letters (2013-08-21)
Lovely Angel Panamparambil Antony, Tomáš Slanina, Peter Šebej, Tomáš Šolomek, Petr Klán
ABSTRACT
6-Hydroxy-3-oxo-3H-xanthene-9-carboxylic acid is introduced as the first transition-metal-free carbon monoxide releasing molecule activated by visible light (photoCORM). This water-soluble fluorescein analogue releases carbon monoxide in both water and methanol upon irradiation at 500 nm. When selectively irradiated in the presence of hemoglobin (Hb) under physiological conditions, released CO is quantitatively trapped to form carboxyhemoglobin (COHb). The reaction progress can be accurately monitored by characteristic absorption and emission properties of the reactants and products.
Sigma-Aldrich
Xanthene-9-carboxylic acid, 98%
Sigma-Aldrich
Fluorescein sodium salt, JIS special grade
Sigma-Aldrich
Fluorescein sodium salt, BioReagent, suitable for fluorescence
Sigma-Aldrich
Fluorescein sodium salt, p.a., for source staining
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer