MilliporeSigma
  • Structural variability between starch granules in wild type and in ae high-amylose mutant maize kernels.
Structural variability between starch granules in wild type and in ae high-amylose mutant maize kernels.
Carbohydrate polymers (2013-08-06)
Dongli Liu, Mary L Parker, Nikolaus Wellner, Andrew R Kirby, Kathryn Cross, Victor J Morris, Fang Cheng
ABSTRACT
Starch granule structure within wild-type and ae high-amylose mutant maize kernels has been mapped in situ using light, electron and atomic force microscopy, and both Raman and infra-red spectroscopy. The population of wild-type starch granules is found to be homogenous. The ae mutant granule population is heterogeneous. Heterogeneity in chemical and physical structure is observed within individual granules, between granules within cells, and spatially within the kernel. The highest level of heterogeneity is observed in the region where starch is first deposited during kernel development. Light microscopy demonstrates structural diversity through use of potassium iodide/iodine staining and polarised microscopy. Electron and atomic force microscopy, and infra-red and Raman spectroscopy defined the nature of the structural changes within granules. The methodology provides novel information on the changes in starch structure resulting from kernel development.
Sigma-Aldrich
Potassium iodide, SAJ first grade, ≥99.5%
Sigma-Aldrich
Potassium iodide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Amylose from potato, used as amylase substrate
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, JIS special grade, ≥99.5%
Sigma-Aldrich
Potassium iodide, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥99.0%
Sigma-Aldrich
Potassium iodide, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium iodide, puriss., meets analytical specification of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium iodide, ReagentPlus®, 99%
Sigma-Aldrich
Gold etchant, standard
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, puriss., ≥99.5% (RT), particles (round)