MilliporeSigma
  • Role of organic cation transporters in the ocular disposition of its intravenously injected substrate in rabbits: implications for ocular drug therapy.
Role of organic cation transporters in the ocular disposition of its intravenously injected substrate in rabbits: implications for ocular drug therapy.
Experimental eye research (2013-07-31)
Jayabalan Nirmal, Anju Sirohiwal, Sundararajan Baskar Singh, Nihar Ranjan Biswas, Vasantha Thavaraj, Raj Vardhan Azad, Thirumurthy Velpandian
ABSTRACT
The present study was conducted to test the hypothesis; OCT may be active from blood-to-vitreous for the uptake of its substrates. Ocular uptake of Tetraethylammonium (TEA) across blood ocular barriers and the tissue distribution was evaluated in vivo in New Zealand albino rabbits after intravenous administration. Quinidine (blocker) pretreatment resulted in a significant (p < 0.05) reduction in the Area Under the Curve (AUC) of TEA in vitreous (4.2 fold) and aqueous humor (1.8 fold) as compared to the control group which supports the role of OCT in uptake transport of its substrate across Blood ocular barrier. The blockade of OCT also affected the elimination of its substrate resulting in increased plasma levels. In most of the tissues, OCT are functionally present from apical to basolateral. The gene expression studies also showed the presence of OCT1, OCTN1 and OCTN2 in various ocular tissues studied. The present findings suggest that OCT are functionally active in blood ocular barriers and involved in the transport of its substrate from blood-to-vitreous humor.
Sigma-Aldrich
Tetraethylammonium chloride hydrate
Sigma-Aldrich
Tetraethylammonium iodide, 98%
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)
Sigma-Aldrich
Tetraethylammonium bromide, ReagentPlus®, 99%
Supelco
Tetraethylammonium chloride, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetraethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium bromide, reagent grade, 98%
Sigma-Aldrich
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)