MilliporeSigma
  • Perovskite LaTiO₃-Ag0.2 nanomaterials for nonenzymatic glucose sensor with high performance.
Perovskite LaTiO₃-Ag0.2 nanomaterials for nonenzymatic glucose sensor with high performance.
Biosensors & bioelectronics (2013-05-08)
Yin-zhu Wang, Hui Zhong, Xiao-mo Li, Fei-fei Jia, Yi-xiang Shi, Wei-guang Zhang, Zhi-peng Cheng, Li-li Zhang, Ji-kui Wang
ABSTRACT
In this paper, a nonenzymatic glucose biosensor based on perovskite LaTiO3-Ag0.2(LTA) modified electrode was presented. The morphology and the composition of the perovskite LaTiO₃-Ag0.2 nanomaterials were characterized by using scanning electron microscopy (SEM) and X-ray diffraction (XRD) respectively. The LaTiO₃-Ag0.2(LTA) composite was investigated by electrochemical characterization using cyclic voltammetry (CV) and electrochemical impedance spectroscopy (EIS). Under optimal conditions, CV and chronoamperometry (I-t) study revealed that, compared with the bare glassy carbon electrode (GCE), the modified electrode showed a remarkable increase in the efficiency of the electrocatalytic oxidation of glucose, starting at around +0.70 V (vs. Ag/AgCl). The prepared sensor exhibited a high sensitivity of 784.14 µAmM⁻¹ cm⁻², a low detection limit of 2.1×10⁻⁷ M and a wide linear range from 2.5 µM to 4 mM (R=0.9997). More importantly, the LTA modified electrode was also relatively insensitive to commonly interfering species such as ascorbic acid (AA), uric acid (UA), dopamine (DA) in high potential. Moreover, the nonenzymatic sensor was applied to the determination of glucose in human serum samples and the results were in good agreement with clinical data. Electrodes modified with perovskite nanomaterials are highly promising for nonenzymatic electrochemical detection of glucose because of their high sensitivity, fast response, excellent stability and good reproducibility.
Sigma-Aldrich
Lanthanum, powder, −40 mesh, under oil, 99.9% trace rare earth metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Lanthanum, pieces
Pricing and availability is not currently available.
Sigma-Aldrich
Calcium titanate, nanopowder, <100 nm particle size (BET), 99% trace metals basis
Pricing and availability is not currently available.