MilliporeSigma
  • Improving the osteogenic potential of BMP-2 with hyaluronic acid hydrogel modified with integrin-specific fibronectin fragment.
Improving the osteogenic potential of BMP-2 with hyaluronic acid hydrogel modified with integrin-specific fibronectin fragment.
Biomaterials (2012-10-30)
Marta Kisiel, Mikaël M Martino, Manuela Ventura, Jeffrey A Hubbell, Jöns Hilborn, Dmitri A Ossipov
ABSTRACT
While human bone morphogenetic protein-2 (rhBMP-2) is a promising growth factor for bone regeneration, its clinical efficacy has recently shown to be below expectation. In order to improve the clinical translation of rhBMP-2, there exists strong motivation to engineer better delivery systems. Hyaluronic acid (HA) hydrogel is a suitable carrier for the delivery of rhBMP-2, but a major limitation of this scaffold is its low cell adhesive properties. In this study, we have determined whether covalent grafting of an integrin-specific ligands into HA hydrogel could improve cell attachment and further enhance the osteogenic potential of rhBMP-2. A structurally stabilized fibronectin (FN) fragment containing the major integrin-binding domain of full-length FN (FN III9*-10) was engineered, in order to be incorporated into HA hydrogel. Compared to non-functionalized HA hydrogel, HA-FN hydrogel remarkably improved the capacity of the material to support mesenchymal stem cell attachment and spreading. In an ectopic bone formation model in the rat, delivery of rhBMP-2 with HA-FN hydrogel resulted in the formation of twice as much bone with better organization of collagen fibers compared to delivering the growth factor in non-functionalized HA hydrogel. This engineered hydrogel carrier for rhBMP-2 can be relevant in clinical bone repair.
Sigma-Aldrich
TissueFab® bioink kit, Fibronectin-UV/365nm
Sigma-Aldrich
Hyaluronic acid sodium salt from Streptococcus equi, mol wt 90,000-110,000