- [Formation of indium-111 in alpha and helium-3 bombardment of natural cadmium (author's transl)].
[Formation of indium-111 in alpha and helium-3 bombardment of natural cadmium (author's transl)].
Radioisotopes (1979-11-01)
Y Nakamura, H Nakahara, Y Murakami
PMID531249
ABSTRACT
Formation cross sections and thick-target yields were studied in detail for alpha reactions and helium-3 reactions of natural cadmium in the energy range 10-40 MeV in order to determine the optimum condition for the production of medical use 11In. The helium-3 reactions were found to give higher 111In yield; namely, at the optimum helium-3 particle energy of 40 MeV and with the cadmium foil of 280 mg/cm2 thickness the amount of 11In produced was 270 muCi/muA . h. A cooling-time of 54 hours was required for the best radiochemical purity. The extraction of HBr-isopropyl ether followed by the anion exchange method was found to be the best chemical method for the carrier-free indium-111 separation.
Product Description
Cadmium, foil, 4mm disks, thickness 0.05mm, as rolled, 99.99+%
Cadmium, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 50x200mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, foil, not light tested, 50x50mm, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 25x25mm, thickness 0.005mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.01mm, 99.7+%
Cadmium, foil, 8mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, not light tested, 100x100mm, thickness 0.005mm, 99.7+%
Cadmium, foil, light tested, 100x100mm, thickness 0.015mm, 99.7+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.01mm, 99.7+%
Cadmium, foil, light tested, 25x25mm, thickness 0.01mm, 99.7+%
Cadmium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 50mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, 8mm disks, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 50mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, not light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 6mm disks, thickness 0.005mm, 99.7+%
Cadmium, foil, not light tested, 100x100mm, thickness 0.025mm, 99.7+%
Cadmium, foil, light tested, 25x25mm, thickness 0.015mm, 99.7+%
Cadmium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, 25mm disks, thickness 0.5mm, as rolled, 99.95%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, foil, 15mm disks, thickness 0.25mm, as rolled, 99.95%
Cadmium, foil, 10mm disks, thickness 0.01mm, 99.7+%
Cadmium, foil, 30x100mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.9%
Cadmium, foil, 1m coil, thickness 0.5mm, as rolled, 100%