MilliporeSigma
  • A class of 2,4-bisanilinopyrimidine Aurora A inhibitors with unusually high selectivity against Aurora B.
A class of 2,4-bisanilinopyrimidine Aurora A inhibitors with unusually high selectivity against Aurora B.
Journal of medicinal chemistry (2009-05-01)
Ignacio Aliagas-Martin, Dan Burdick, Laura Corson, Jennafer Dotson, Jason Drummond, Carter Fields, Oscar W Huang, Thomas Hunsaker, Tracy Kleinheinz, Elaine Krueger, Jun Liang, John Moffat, Gail Phillips, Rebecca Pulk, Thomas E Rawson, Mark Ultsch, Leslie Walker, Christian Wiesmann, Birong Zhang, Bing-Yan Zhu, Andrea G Cochran
ABSTRACT
The two major Aurora kinases carry out critical functions at distinct mitotic stages. Selective inhibitors of these kinases, as well as pan-Aurora inhibitors, show antitumor efficacy and are now under clinical investigation. However, the ATP-binding sites of Aurora A and Aurora B are virtually identical, and the structural basis for selective inhibition has therefore not been clear. We report here a class of bisanilinopyrimidine Aurora A inhibitors with excellent selectivity for Aurora A over Aurora B, both in enzymatic assays and in cellular phenotypic assays. Crystal structures of two of the inhibitors in complex with Aurora A implicate a single amino acid difference in Aurora B as responsible for poor inhibitory activity against this enzyme. Mutation of this residue in Aurora B (E161T) or Aurora A (T217E) is sufficient to swap the inhibition profile, suggesting that this difference might be exploited more generally to achieve high selectivity for Aurora A.
Sigma-Aldrich
2,4-Dichloro-5-fluoropyrimidine, 97%