MilliporeSigma
  • Tröger's base-functionalised organic nanoporous polymer for heterogeneous catalysis.
Tröger's base-functionalised organic nanoporous polymer for heterogeneous catalysis.
Chemical communications (Cambridge, England) (2010-01-29)
Xin Du, Yalei Sun, Bien Tan, Qingfeng Teng, Xiaojun Yao, Chengyong Su, Wei Wang
ABSTRACT
An organic nanoporous polymer (BET surface area of 750 m(2) g(-1)) containing bicovalently-bonded Tröger's base as functional moieties in the network was synthesized, characterised, and further applied as a promising heterogeneous catalyst in the addition reaction of diethylzinc to aromatic aldehyde.
Product Description
Troger′s Base, 98%