MilliporeSigma
  • Actinide-mediated coupling of 4-fluorobenzonitrile: synthesis of an eight-membered thorium(IV) tetraazametallacycle.
Actinide-mediated coupling of 4-fluorobenzonitrile: synthesis of an eight-membered thorium(IV) tetraazametallacycle.
Chemical communications (Cambridge, England) (2007-02-28)
Eric J Schelter, David E Morris, Brian L Scott, Jaqueline L Kiplinger
ABSTRACT
An eight-membered thorium(IV) tetraazamacrocycle is produced by the sequential, metal-mediated coupling of four equivalents of 4-fluorobenzonitrile; its formation is consistent with the involvement of an imido intermediate, generated from a thorium ketimide complex.
Sigma-Aldrich
4-Fluorobenzonitrile, 99%